Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ XE 991 dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective blocker of KV7 (KCNQ) voltage-gated potassium channels. Blocks KV7.2+7.3 (KCNQ2+3)/ M-currents (IC50 = 0.6 - 0.98 μM) and KV7.1 (KCNQ1) homomeric channels (IC50 = 0.75 μM) but is less potent against KV7.1/minK channels (IC50 = 11.1 μM). Augments hippocampal ACh release and is a cognitive enhancer following oral administration in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 449.37 |
| Purity | >99% |
| Molecular Weight (g/mol) | 449.375 |
| Chemical Name or Material | XE 991 dihydrochloride |
Safety and Handling
| Recommended Storage | Desiccate at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction