Learn More
XRCC1 Polyclonal Antibody, Rockland™

Chicken Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | XRCC1 |
---|---|
Concentration | 0.93 mg/mL |
Applications | ELISA, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1298
![]() ![]() |
Rockland Immunochemicals
200901MG5 |
100 μg |
Each for $636.24
|
|
|||||
50-199-1299
![]() ![]() |
Rockland Immunochemicals
200901MG5S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This purified antibody is directed against human XRCC1. This product is an IgY fraction antibody purified from monospecific chicken egg yolks by a multi-step process which includes selective precipitation and salt fractionation followed by extensive dialysis against the buffer stated above.
XRCC1 (X-ray cross complementing factor-1) is involved in DNA base excision repair. XRCC1 is a 70 kDa protein that has been shown to be physically associated with other DNA repair enzymes, including poly (ADP-ribose) polymerase (PARP), DNA Ligase III and DNA polymerase beta. XRCC1 contains a BRCT domain (for BRCA1 carboxyl terminus) which was originally found in the tumor suppressor protein BRCA1. Recently, the BRCT domain has been shown to mediate the protein-protein interaction of XRCC1 and DNA ligase III-alpha.Specifications
XRCC1 | |
ELISA, Immunofluorescence, Western Blot | |
Unconjugated | |
Chicken | |
Human, Mouse | |
P18887, Q60596 | |
22594, 7515 | |
Anti-XRCC1 antibody was prepared from eggs of chickens laid after repeated immunizations with a synthetic peptide corresponding to a N-Terminal portion of human XRCC1 conjugated to Keyhole Limpet Hemocyanin (KLH). | |
-20°C, Avoid Freeze/Thaw Cycles | |
XRCC1 |
0.93 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.002M sodium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7 | |
DNA repair protein XRCC1; RCC; X-ray repair complementing defective repair in Chinese hamster cells 1; X-ray repair cross complementing 1; x-ray repair cross-complementing group 1 protein; X-ray repair cross-complementing protein 1; Xrcc1; Xrcc-1 | |
XRCC1 | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.