Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ YC 1

Click to view available options
Quantity:
10 mg
50 mg
Description
Nitric oxide-independent activator of soluble guanylyl cyclase (sGC). Significantly elevates cGMP levels and inhibits collagen-stimulated aggregation of washed rabbit platelets (IC50 = 14.6 μM); induces relaxation in denuded phenylephrine-contracted rabbit aortic rings (EC50 = 1.9 μM). Also displays antiproliferative activity in vitro and in vivo by inducing G1 cell cycle arrest in two human hepatocellular carcinoma (HCC) cell lines, and in HCC xenografts in athymic SCID mice. Exhibits low cytotoxicity in non-malignant cells.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 304.34 |
| Purity | >99% |
| Molecular Weight (g/mol) | 304.35 |
| Chemical Name or Material | YC 1 |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction