Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Z-DNA Antibody, Novus Biologicals™
SDP

Catalog No. p-7200937 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.125 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017818 0.025 mg
NB100749 0.125 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017818 Supplier Novus Biologicals Supplier No. NB1007490.025MG

Sheep Polyclonal Antibody has been used in 2 publications

Z-DNA Polyclonal specifically detects Z-DNA in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Z-DNA
Applications Immunohistochemistry (Paraffin), Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration 2.1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10, Immunoprecipitation 8 ug/ml, Immunohistochemistry-Paraffin 1:10
Formulation PBS (pH 7.5) with No Preservative
Host Species Sheep
Immunogen Immunogen was brominated poly-(DG-DC)-left handed DNA (Z-DNA) complexed to methylated BSA
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.