Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC18 Antibody, Novus Biologicals™
SDP

Catalog No. p-7108482 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP160116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP160116 Supplier Novus Biologicals Supplier No. NBP160116
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZDHHC18 Polyclonal specifically detects ZDHHC18 in Human samples. It is validated for Western Blot.

Specifications

Antigen ZDHHC18
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9NUE0
Gene Alias DHHC domain containing 18, DHHC18, DHHC-18, EC 2.3.1, EC 2.3.1.-, zinc finger, DHHC-type containing 18
Gene Symbols ZDHHC18
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ZDHHC18 (zinc finger, DHHC-type containing 18) The peptide sequence was selected from the middle region of ZDHHC18. Peptide sequence FSIWSILGLSGFHTYLVASNLTTNEDIKGSWSSKRGGEASVNPYSHKSII.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84243
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.