accessibility menu, dialog, popup

UserName

Viewing profile as user:
SHNNOFFF

CAS RN 284028-89-3

IUPAC Name: 2-[4-(trifluoromethyl)phenyl]-1H,4H,5H,7H,8H-thiopyrano[4,3-d]pyrimidin-4-one
Synonyms: 2-[4-(trifluoromethyl)phenyl]-1H,5H,7H,8H-thiopyrano[4,3-d]pyrimidin-4-one
Molecular Weight (g/mol): 312.31
Molecular Formula: C14H11F3N2OS
InChi Key: KLGQSVMIPOVQAX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)C1=NC(=O)C2=C(CCSC2)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results