CAS RN 5996-10-1
CAS RN 5996-10-1
IUPAC Name:
(2S,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol hydrate
Synonyms:
α-glucose hydrate
Molecular Weight (g/mol):
198.17
Molecular Formula:
C6H14O7
InChi Key:
OSNSWKAZFASRNG-WNFIKIDCSA-N
SMILES:
O.OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O
1
–
1
of
1
results
1
Dextrose, Monohydrate, Multi-Compendial, U.S.P., J.T. Baker™
CAS: 5996-10-1 Molecular Formula: C6H14O7 Molecular Weight (g/mol): 198.17 InChI Key: OSNSWKAZFASRNG-WNFIKIDCSA-N Synonym: 6-hydroxymethyl oxane-2,3,4,5-tetrol hydrate,d-galactose hydrate,dextrose monohydrate, usp,.alpha.-d-glucose hydrate,dextrose, u.s.p. PubChem CID: 133126654 IUPAC Name: (2R,3S,4R,5R,6S)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol;hydrate SMILES: C(C1C(C(C(C(O1)O)O)O)O)O.O