CAS RN 7048-04-6
CAS RN 7048-04-6
Thermo Scientific Chemicals L-Cysteine hydrochloride monohydrate, 99%
CAS: 7048-04-6 Molecular Formula: C3H10ClNO3S Molecular Weight (g/mol): 175.63 MDL Number: MFCD00065606 InChI Key: QIJRTFXNRTXDIP-JIZZDEOASA-N Synonym: l-cysteine hydrochloride monohydrate,h-cys-oh.hcl.h2o,l-cysteine hydrochloride hydrate,l-cysteine monohydrate monochloride,r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate,unii-zt934n0x4w,l-cysteine hydrate hydrochloride,cysteine hydrochloride monohydrate, l,h-cys-ohhclh2o,cysteine hcl PubChem CID: 23462 IUPAC Name: (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride SMILES: C(C(C(=O)O)N)S.O.Cl
Fisher Chemical L-Cysteine Hydrochloride Monohydrate (White Crystals), Fisher BioReagents

CAS: 7048-04-6 Molecular Formula: C3H10ClNO3S Molecular Weight (g/mol): 175.63 InChI Key: QIJRTFXNRTXDIP-JIZZDEOASA-N Synonym: l-cysteine hydrochloride monohydrate,h-cys-oh.hcl.h2o,l-cysteine hydrochloride hydrate,l-cysteine monohydrate monochloride,r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate,unii-zt934n0x4w,l-cysteine hydrate hydrochloride,cysteine hydrochloride monohydrate, l,h-cys-ohhclh2o,cysteine hcl PubChem CID: 23462 IUPAC Name: (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride SMILES: C(C(C(=O)O)N)S.O.Cl
MP Biomedicals, Inc L-Cysteine hydrochloride monohydrate, MP Biomedicals™
CAS: 7048-04-6 Molecular Formula: C3H10ClNO3S Molecular Weight (g/mol): 175.63 MDL Number: MFCD00065606 InChI Key: QIJRTFXNRTXDIP-JIZZDEOASA-N Synonym: l-cysteine hydrochloride monohydrate,h-cys-oh.hcl.h2o,l-cysteine hydrochloride hydrate,l-cysteine monohydrate monochloride,r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate,unii-zt934n0x4w,l-cysteine hydrate hydrochloride,cysteine hydrochloride monohydrate, l,h-cys-ohhclh2o,cysteine hcl PubChem CID: 23462 IUPAC Name: (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride SMILES: C(C(C(=O)O)N)S.O.Cl
MP Biomedicals, Inc L-Cysteine Hydrochloride Monohydrate, MP Biomedicals
Molecular Formula: C3H10ClNO3S Molecular Weight (g/mol): 175.63 MDL Number: MFCD00065606 InChI Key: QIJRTFXNRTXDIP-JIZZDEOASA-N
Thermo Scientific L-Cysteine Hydrochloride Monohydrate, 98.5 to 101.5% (Dry Basis)
CAS: 7048-04-6 Molecular Formula: C3H10ClNO3S Molecular Weight (g/mol): 175.63 MDL Number: MFCD00065606 InChI Key: QIJRTFXNRTXDIP-JIZZDEOASA-N Synonym: l-cysteine hydrochloride monohydrate,h-cys-oh.hcl.H2O,l-cysteine hydrochloride hydrate,l-cysteine monohydrate monochloride,r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate,unii-zt934n0x4w,l-cysteine hydrate hydrochloride,cysteine hydrochloride monohydrate, l,h-cys-ohhclh2o,cysteine hcl PubChem CID: 23462 IUPAC Name: (2 R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride SMILES: C(C(C(=O)O)N)S.O.Cl
Spectrum Chemical Manufacturing Corporation L-Cysteine Monohydrochloride, Monohydrate, FCC, 98-101.5%, Spectrum™ Chemical

CAS: 7048-04-6 Molecular Formula: C3H10ClNO3S Molecular Weight (g/mol): 175.63 InChI Key: QIJRTFXNRTXDIP-JIZZDEOASA-N IUPAC Name: hydrogen (2R)-2-amino-3-sulfanylpropanoic acid hydrate chloride SMILES: [H+].O.[Cl-].N[C@@H](CS)C(O)=O
Spectrum Chemical Manufacturing Corporation Cysteine Hydrochloride, Monohydrate, USP, 98.5-101.5%, Spectrum™ Chemical

CAS: 7048-04-6 Molecular Formula: C3H10ClNO3S Molecular Weight (g/mol): 175.63 InChI Key: QIJRTFXNRTXDIP-JIZZDEOASA-N IUPAC Name: hydrogen (2R)-2-amino-3-sulfanylpropanoic acid hydrate chloride SMILES: [H+].O.[Cl-].N[C@@H](CS)C(O)=O
Advanced Chem Tech H-Cys-OH.HCl.H2O, Advanced ChemTech
CAS: 7048-04-6 Molecular Formula: C3H10ClNO3S Molecular Weight (g/mol): 175.63 InChI Key: QIJRTFXNRTXDIP-JIZZDEOASA-N IUPAC Name: hydrogen (2R)-2-amino-3-sulfanylpropanoic acid hydrate chloride SMILES: [H+].O.[Cl-].N[C@@H](CS)C(O)=O
Thermo Scientific Production Chemicals and Services L-Cysteine HCL H2O USP/EP/FCC, Ajinomoto
CAS: 7048-04-6 Molecular Weight (g/mol): 175.63
Avantor J.T.Baker L-Cysteine Hydrochloride, Monohydrate, Multi-Compendial, U.S.P., J.T. Baker™
CAS: 7048-04-6 Molecular Formula: C3H10ClNO3S Molecular Weight (g/mol): 175.63 InChI Key: QIJRTFXNRTXDIP-JIZZDEOASA-N Synonym: l-cysteine hydrochloride monohydrate,h-cys-oh.hcl.h2o,l-cysteine hydrochloride hydrate,l-cysteine monohydrate monochloride,r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate,unii-zt934n0x4w,l-cysteine hydrate hydrochloride,cysteine hydrochloride monohydrate, l,h-cys-ohhclh2o,cysteine hcl PubChem CID: 23462 IUPAC Name: (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride SMILES: C(C(C(=O)O)N)S.O.Cl