Thermometers and Temperature Measurement
Devices for measuring and controlling temperature in reagents, equipment, and reaction vessels; includes digital- and analog-display thermometers, temperature controllers, temperature indicator tapes, and thermometer accessories.
Select from an assortment of thermometers, including traditional mercury thermometers, precision digital thermometers, electronic thermometers, and infrared thermometers.
Digital thermometers can cater to a wide range of projects in both clinical labs and classrooms. Choose from a wide selection of digital infrared thermometers available in both pen and handheld styles for taking readings through tissue, as well as laser thermometers for taking readings from a distance.
Many thermometers, including the affordable student thermometer, can be used for both educational and general applications, while others are perfect for medical applications.
Filtered Search Results
Fisherbrand™ General Purpose Liquid-in-Glass Partial Immersion Thermometers
Fisherbrand™ General Purpose Liquid-in-Glass Partial Immersion Thermometers are ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories and the pharmaceutical industry.
| Thermometer Fill | Blue Organic Liquid |
|---|---|
| Immersion Type | Partial Immersion |
| Material | Glass |
| Certifications/Compliance | ISO/IEC 17025:2017, A2LA, NIST |
| For Use With (Application) | Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries |
Fisherbrand™ Traceable™ Hi-Accuracy Refrigerator Thermometer
Traceable™ Hi-Accuracy Refrigerator Thermometer can monitor two different areas simultaneously. Has the ability to monitor temperatures in refrigerator and freezer simultaneously or two refrigerator locations.
| Width (English) | 2.75 in. |
|---|---|
| Length (Metric) Cable | 304.8 cm |
| Length (Metric) | 114 mm |
| Length (English) | 4.5 in. |
| Min. Temperature (English) | -58°F |
| For Use With (Application) | Monitoring temperature in refrigerator and freezer simultaneously or two refrigerator locations |
| Width (Metric) | 6.9 cm |
| Accuracy | ±0.3°C |
| For Use With (Equipment) | Refrigerators, Freezers |
| Max. Temperature (English) | 158°F |
| Length (English) Cable | 120 in. |
| Certifications/Compliance | ISO 17025, A2LA, NIST |
| Min. Temperature (Metric) | -50°C |
| Calibrated | Yes |
| Weight (Metric) | 140 g |
| Diameter (Metric) Probe | 4.57 mm |
| Material | ABS Plastic |
| Length (English) Probe | 0.7 in. |
| Max. Temperature (Metric) | 70°C |
| Length (Metric) Probe | 1.9 cm |
| Product Type | Digital Thermometer |
| Diameter (English) Probe | 0.18 in. |
| Temperature Range (English) | -58°F to +158°F |
| Weight (English) | 5 oz. |
| Temperature Range (Metric) | -50°C to +70°C |
| Display | LED |
| Resolution | 0.01°C |
Fisherbrand™ Traceable™ Refrigerator/Freezer Ultra™ Thermometer
Science Education
A science education product.
A science education product.
Monitor two different areas at the same time with Traceable™ Refrigerator/Freezer Ultra™ Thermometer.
| Bottle Size | 2.5 x 6.3 cm (1 x 2.5 in.) |
|---|---|
| Min. Temperature (English) | -58°F |
| Max. Temperature (Metric) | 70°C |
| For Use With (Application) | Monitoring temperatures in freezers, water baths, heating blocks, incubators, and refrigerators |
| Width (Metric) | 6.99 cm |
| Accuracy | ±0.5°C (-30°C to +50°C), ±1°C otherwise |
| For Use With (Equipment) | Monitoring Temperature in Freezer, Water Bath, Heating Block, Incubator and Refrigerator |
| Product Type | Refrigerator/Freezer Ultra Thermometer |
| Max. Temperature (English) | 158°F |
| Temperature Range (English) | -58°F to +158°F |
| Weight (English) | 4 oz. |
| Temperature Range (Metric) | -50°C to +70°C |
| Min. Temperature (Metric) | -50°C |
| Calibrated | Yes |
| Display | Digital |
| Resolution | 0.1° |
Fisherbrand™ Traceable™ International Standards Extreme-Accuracy Digital Thermometer
Traceable™ International Standards Extreme-Accuracy Digital Thermometers ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater.
| Height (Metric) | 1.9 cm |
|---|---|
| Width (English) | 2.75 in. |
| Length (Metric) | 107 mm |
| Length (English) | 4.25 in. |
| Material | ABS Plastic |
| Length (English) Probe | 6.25 in. |
| For Use With (Application) | Measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater |
| Width (Metric) | 6.9 cm |
| Battery Type | AA |
| Length (Metric) Probe | 15.9 cm |
| Includes | Stainless Steel probe, High-impact, chemical-resistant ABS plastic case, stand, Velcro™, magnetic strips, wall mount, two AA batteries (allow continuous, 1-year always-on monitoring) |
| Dimensions (L x W x H) | 4.25 x 2.75 x 0.75 in. (10.7 x 6.9 x 1.9 cm) |
| For Use With (Equipment) | Water Baths |
| Product Type | Digital Thermometer |
| Weight (English) | 5 oz. |
| Certifications/Compliance | ISO 17025, A2LA, NIST |
| Calibrated | Yes |
| Display | LED |
| Weight (Metric) | 140 g |
| Resolution | 0.01°C |
Fisherbrand™ Traceable™ Total-Range Thermometer
Technologically advanced, pocket-size thermometer with water-resistant case
Fisherbrand™ Traceable™ Digital Thermometer with Stainless-Steel Stem and 0.25 in. LCD Screen
Traceable™ Digital Thermometer used to measure temperature of gases, food, and semisolids.
| Product Type | Digital Thermometer |
|---|---|
| Min. Temperature (English) | -58°F |
| Material | Stainless Steel |
| Min. Temperature (Metric) | -50°C |
| Calibrated | Yes |
| Display | Digital LCD |
Fisherbrand™ Nonmercury Verification Thermometers
To verify sample temperatures in freezers, refrigerators, and incubators with traceability to NIST.
| Product Type | Verification Thermometer |
|---|---|
| Material | Glass |
| Certifications/Compliance | ISO/IEC 17025:2005, A2LA, NIST |
Fisherbrand™ Precision Liquid-in-Glass Partial and Total Immersion Thermometers
The Fisherbrand™ Precision Liquid-in-Glass Partial and Total Immersion Thermometer is ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories and the pharmaceutical industry.
| Thermometer Fill | Organic |
|---|---|
| Material | Glass |
| Grade | Precision |
| For Use With (Application) | Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries |
Fisherbrand™ PFA Safety Coated Liquid-In-Glass Partial Immersion Thermometers
In the event of breakage, the PFA fluoropolymer safety coating keeps the liquid and glass encapsulated.
| Thermometer Fill | Organic |
|---|---|
| Immersion Type | Partial Immersion |
| Material | Glass |
| Certifications/Compliance | ISO/IEC 17025:2005, A2LA, NIST |
| For Use With (Application) | Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries |
Fisherbrand™ Digital Thermometers with Stainless-Steel Probe on Cable
Easy-to-read digital thermometers are compact to hold in your hand or mount on wall in restricted areas
| Product Type | Traceable Digital Thermometer w/Probe |
|---|---|
| Certifications/Compliance | ISO 17025, A2LA, NIST |
| Calibrated | Yes |
| Display | Digital LCD |
| Length (Metric) Probe | 14.6 cm |
|---|---|
| Length (Metric) Cable | 304.8 cm |
| For Use With (Equipment) | Freezer, incubator, water bath |
| Diameter (English) Probe | 0.14 in. |
| Depth (Metric) | 1.4 cm |
| Length (English) Cable | 120 in. |
| Length (English) Probe | 5.75 in. |
| Certifications/Compliance | ISO 17025, A2LA, NIST |
| Width (Metric) | 8 cm |
| Calibrated | Yes |
| Display | Digital LCD |
Fisherbrand™ Nonmercury Verification Thermometers
To verify sample temperatures in freezers, refrigerators, incubators and ovens with traceability to NIST.
| Product Type | Verification Thermometer |
|---|---|
| Material | Glass |
| Certifications/Compliance | NIST |
Fisherbrand™ Traceable™ Dual Thermometer with Min/Max and Time/Date
Displays date/time when minimum/maximum temperatures occur
| Height (Metric) | 2.5 cm |
|---|---|
| Width (English) | 1.75 in. |
| Length (Metric) Cable | 106.68 cm |
| Length (Metric) | 89 mm |
| Length (English) | 3.5 in. |
| Min. Temperature (English) | -58°F |
| Material | ABS Plastic |
| Max. Temperature (Metric) | 70°C |
| For Use With (Application) | Cold storage, chemicals, pharmaceuticals |
| Width (Metric) | 4.4 cm |
| Accuracy | ±1°C |
| Dimensions (L x W x H) | 3.5 x 1.75 x 1 in. (8.9 x 4.4 x 2.5 cm) |
| Product Type | Digital Thermometer |
| Max. Temperature (English) | 158°F |
| Temperature Range (English) | -58°F to +158°F |
| Length (English) Cable | 42 in. |
| Weight (English) | 3.6 oz. |
| Certifications/Compliance | ISO 17025, A2LA, NIST |
| Temperature Range (Metric) | -50°C to +70°C |
| Min. Temperature (Metric) | -50°C |
| Calibrated | Yes |
| Display | Digital LCD |
| Weight (Metric) | 100 g |
| Resolution | 0.1°C |
| Height (English) | 1 in. |
Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer
Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor.
| Height (Metric) | 19 mm |
|---|---|
| Length (Metric) Cable | 25.4 cm |
| Length (Metric) | 70 mm |
| Length (English) | 2.75 in. |
| Min. Temperature (English) | -58°F |
| Material | ABS Plastic |
| Bottle Fill | Glycol |
| Max. Temperature (Metric) | 70°C |
| For Use With (Application) | Monitoring temperature in freezers, water baths, heating blocks, incubators and refrigerators |
| Battery Type | AA Alkaline (1) |
| Accuracy | ±1°C |
| For Use With (Equipment) | Refrigerators/Freezers |
| Product Type | Fridge/Freezer Digital Thermometer |
| Max. Temperature (English) | 158°F |
| Temperature Range (English) | -58°F to +158°F |
| Length (English) Cable | 10 in. |
| Weight (English) | 4 oz. |
| Certifications/Compliance | ISO 17025, A2LA, NIST |
| Temperature Range (Metric) | -50°C to +70°C |
| Min. Temperature (Metric) | -50°C |
| Calibrated | Yes |
| Display | Digital LCD |
| Resolution | 1°C |
| Height (English) | 0.75 in. |
Fisherbrand™ Traceable™ RTD Platinum Thermometer
High-precision, durable thermometer with platinum RTD probe, and linear fuzzy logic chip for wide temperature-range operation