Thermo Scientific Water Analysis Products and Special Offers
Sample Protection as Top Priority
Thermo Scientific Heratherm General Protocol Ovens
Efficient, safe and easy, Thermo Scientific™ Heratherm™ gravity convection ovens feature an environmentally friendly design with cost savings in mind, reducing energy consumption by as much as 25% over previous models. From simple glassware drying to the most complex and carefully controlled heating applications, the Thermo Scientific Heratherm oven range includes an array of solutions to match your needs.
Catalog No. | Type | Capacity | Footprint (m2 / sq.ft.) | Buy Now | |
---|---|---|---|---|---|
General Protocol Oven | 65L | 0.3 / 3.2 | Shop Now | ||
General Protocol Oven | 105L | 0.36 / 3.9 | Shop Now | ||
General Protocol Oven | 180L | 0.47 / 5.1 | Shop Now |
Turn the Heat Up on Efficiency
Thermo Scientific Thermolyne and Moldatherm Furnaces
Thermo Scientific™ Thermolyne™ muffle furnaces feature fast heatup and excellent energy efficiency in the small benchtop models, or a more rugged design and multiple safety features in the industrial benchtop models. Thermo Scientific™ Lindberg/Blue M™ Moldatherm™ box furnaces include unique insulation and heating element composites, and minimize outer surface temperatures while maintaining uniform heat distribution within the chamber.
Catalog No. | Type | Capacity | Power Consumption | Buy Now | |
---|---|---|---|---|---|
Benchtop Muffle Furnace | 1.3L | 1060w | Shop Now | ||
Benchtop Muffle Furnace | 2.1L | 1450w | Shop Now | ||
Industrial Benchtop Muffle Furnaces | 2.2L | 2230w | Shop Now | ||
Box Furnace | 1.99L | 1800w | Shop Now |
Intelligent Design for Improved Results
Thermo Scientific Heratherm General Protocol Microbiological Incubators
Ideally suited for routine applications in pharmaceutical, medical, food and research laboratories, Thermo Scientific Heratherm general protocol microbiological incubators offer gravity convection for gentle airflow, resulting in minimized drying out of samples. The best choices for applications with open plates or open containers, these incubators keep sample safety in mind.
Catalog No. | Type | Volume (Metric) Chamber | Amperage | Buy Now | |
---|---|---|---|---|---|
General Protocol Microbiological Incubator | 75L | 2.5A | Shop Now | ||
General Protocol Microbiological Incubator | 117L | 4.5A | Shop Now |
Mix-and-Match Your Application and Vessel
Thermo Scientific LP Vortex Mixer
Ideal for vortex mixing in a tube, plate or glass vessel, Thermo Scientific low-profile vortex mixers offer flexibility in application and vessel use, and can help you maximize your lab's efficiency through reliable precision.
Correct Measurements Every Time
Thermo Scientific Orion pH Buffer Bottles, Packs and Single-Use Pouches
Perform accurate, reproducible pH calibrations with color-coded Thermo Scientific™ Orion™ pH buffer bottles, packs and single-use pouches. All Orion pH buffers are traceable to NIST™ standard reference materials. A wide variety of sizes are available from 15mL single-use pouches to large-volume 19L Cubitainers™ and multi-pack sets.
Catalog No. | Type | pH | Quantity | Shop Now |
---|---|---|---|---|
pH Buffer Bottles | 1.68 (Colorless) | 475mL | Shop Now | |
pH Buffer Bottles | 4.01 (Pink) | 475mL | Shop Now | |
pH Buffer Bottles | 5.00 (Orange) | 475mL | Shop Now | |
pH Buffer Bottles | 6.68 (Colorless) | 475mL | Shop Now | |
pH Buffer Bottles | 7.00 (Yellow) | 475mL | Shop Now | |
pH Buffer Bottles | 10.01 (Blue) | 475mL | Shop Now | |
pH Buffer Bottles Pack | 4.01 (Pink) | 5 x 60mL | Shop Now | |
pH Buffer Bottles Pack | 7.00 (Yellow) | 5 x 60mL | Shop Now | |
pH Buffer Individual Use Pouches | 4.01 | 25 x 15mL | Shop Now | |
pH Buffer Individual Use Pouches | 7.00 | 10 x 15mL | Shop Now | |
pH Buffer Individual Use Pouches | 7.00 | 25 x 15mL | Shop Now | |
pH Buffer Individual Use Pouches | 10.01 | 10 x 15mL | Shop Now |
Thermo Scientific Orion pH Electrode Cleaning and Storage Solutions
Restore proper electrode performance and prolong electrode life using Thermo Scientific Orion pH electrode cleaning and storage solutions in ready-to-use bottles.