Assay Details
Gene Family ID: | MIPF0000007, mir-181 |
Mature miRNA Sequence: | AACAUUCAACCUGUCGGUGAGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-181c-5p | MIMAT0000258 | |
| Mouse | mmu-miR-181c-5p | MIMAT0000674 | |
| Rat | rno-miR-181c-5p | MIMAT0000857 | |
| Callithrix jacchus | cja-miR-181c | MIMAT0039344 | |
| Cricetulus griseus | cgr-miR-181c-5p | MIMAT0023801 | |
| Daubentonia madagascariensis | dma-miR-181c | MIMAT0049296 | |
| View More Rows | |||
| Gorilla gorilla | ggo-miR-181c | MIMAT0002511 | |
| Pan paniscus | ppa-miR-181c | MIMAT0002512 | |
| Pan troglodytes | ptr-miR-181c | MIMAT0002510 | |
| Pig | ssc-miR-181c | MIMAT0002144 | |
| Pongo pygmaeus | ppy-miR-181c | MIMAT0015778 | |
| Rhesus monkey | mml-miR-181c-5p | MIMAT0002509 | |
| Tupaia chinensis | tch-miR-181c-5p | MIMAT0036526 | |
Stem-loop Details
Human
Mouse
Rat
Callithrix jacchus
Cricetulus griseus
Daubentonia madagascariensis
Gorilla gorilla
Pan paniscus
Pan troglodytes
Pig
Pongo pygmaeus
Rhesus monkey
Tupaia chinensis
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products
miRBase Alias:
Human: hsa-miR-181c (v17)
Mouse: mmu-miR-181c (v17)
Rat: rno-miR-181c (v18)
Macaca mulatta: mml-miR-181c (v19)
Mouse: mmu-miR-181c (v17)
Rat: rno-miR-181c (v18)
Macaca mulatta: mml-miR-181c (v19)

Genomic Map