Assay Details
Gene Family ID: | MIPF0000007, mir-181 |
Mature miRNA Sequence: | AACAUUCAACCUGUCGGUGAGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-181c-5p | MIMAT0000258 |
| Mouse | mmu-miR-181c-5p | MIMAT0000674 |
| Rat | rno-miR-181c-5p | MIMAT0000857 |
| Callithrix jacchus | cja-miR-181c | MIMAT0039344 |
| Cricetulus griseus | cgr-miR-181c-5p | MIMAT0023801 |
| Daubentonia madagascariensis | dma-miR-181c | MIMAT0049296 |
| View More Rows | ||
| Gorilla gorilla | ggo-miR-181c | MIMAT0002511 |
| Pan paniscus | ppa-miR-181c | MIMAT0002512 |
| Pan troglodytes | ptr-miR-181c | MIMAT0002510 |
| Pig | ssc-miR-181c | MIMAT0002144 |
| Pongo pygmaeus | ppy-miR-181c | MIMAT0015778 |
| Rhesus monkey | mml-miR-181c-5p | MIMAT0002509 |
| Tupaia chinensis | tch-miR-181c-5p | MIMAT0036526 |
Stem-loop Details
Human
Mouse
Rat
Callithrix jacchus
Cricetulus griseus
Daubentonia madagascariensis
Gorilla gorilla
Pan paniscus
Pan troglodytes
Pig
Pongo pygmaeus
Rhesus monkey
Tupaia chinensis

Genomic Map