Assay Details
Gene Family ID: | MIPF0000007, mir-181 |
Mature miRNA Sequence: | AACAUUCAACCUGUCGGUGAGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-181c-5p | MIMAT0000258 |
Mouse | mmu-miR-181c-5p | MIMAT0000674 |
Rat | rno-miR-181c-5p | MIMAT0000857 |
Callithrix jacchus | cja-miR-181c | MIMAT0039344 |
Cricetulus griseus | cgr-miR-181c-5p | MIMAT0023801 |
Daubentonia madagascariensis | dma-miR-181c | MIMAT0049296 |
View More Rows | ||
Gorilla gorilla | ggo-miR-181c | MIMAT0002511 |
Pan paniscus | ppa-miR-181c | MIMAT0002512 |
Pan troglodytes | ptr-miR-181c | MIMAT0002510 |
Pig | ssc-miR-181c | MIMAT0002144 |
Pongo pygmaeus | ppy-miR-181c | MIMAT0015778 |
Rhesus monkey | mml-miR-181c-5p | MIMAT0002509 |
Tupaia chinensis | tch-miR-181c-5p | MIMAT0036526 |
Stem-loop Details
Human
Mouse
Rat
Callithrix jacchus
Cricetulus griseus
Daubentonia madagascariensis
Gorilla gorilla
Pan paniscus
Pan troglodytes
Pig
Pongo pygmaeus
Rhesus monkey
Tupaia chinensis
