Assay Details
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | UGUAAACAUCCCCGACUGGAAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-30d-5p | MIMAT0000245 |
| Mouse | mmu-miR-30d-5p | MIMAT0000515 |
| Rat | rno-miR-30d-5p | MIMAT0000807 |
| Anolis carolinensis | aca-miR-30d-5p | MIMAT0021926 |
| Chicken | gga-miR-30d | MIMAT0001129 |
| Fugu rubripes | fru-miR-30d | MIMAT0002949 |
| View More Rows | ||
| Gorilla gorilla | ggo-miR-30d | MIMAT0002618 |
| Horse | eca-miR-30d | MIMAT0013006 |
| Macaca nemestrina | mne-miR-30d | MIMAT0002619 |
| Monodelphis domestica | mdo-miR-30e-5p | MIMAT0050218 |
| Pan paniscus | ppa-miR-30d | MIMAT0002620 |
| Pan troglodytes | ptr-miR-30d | MIMAT0002617 |
| Pongo pygmaeus | ppy-miR-30d | MIMAT0015738 |
| Rhesus monkey | mml-miR-30d-5p | MIMAT0006171 |
| Tetraodon nigroviridis | tni-miR-30d | MIMAT0002950 |
| Xenopus tropicalis | xtr-miR-30d | MIMAT0003657 |
| Zebrafish | dre-miR-30d | MIMAT0001806 |
Stem-loop Details
Human
Mouse
Rat
Anolis carolinensis
Chicken
Fugu rubripes
Gorilla gorilla
Horse
Macaca nemestrina
Monodelphis domestica
Pan paniscus
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
Tetraodon nigroviridis
Xenopus tropicalis
Zebrafish
More Information
Related Products
miRBase Alias:
Human: hsa-miR-30d (v17)
Mouse: mmu-miR-30d (v17)
Rat: rno-miR-30d (v18)
Aeromonas caviae Ae398: aca-miR-30d (v18)
Macaca mulatta: mml-miR-30d (v19)
Mouse: mmu-miR-30d (v17)
Rat: rno-miR-30d (v18)
Aeromonas caviae Ae398: aca-miR-30d (v18)
Macaca mulatta: mml-miR-30d (v19)

Genomic Map