Assay Details
Amplicon Length: | 89 |
Chromosome Location: | Chr. 7 - 109286110 - 109286110 [+] on 109286110 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-30d-5p | MIMAT0000807 |
Rat | rno-miR-30d-3p | MIMAT0004722 |
Stem-loop Details
Rat
