Assay Details
Amplicon Length: | 75 |
Chromosome Location: | Chr. 15 - 68340904 - 68340904 [+] on 68340904 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-30d-5p | MIMAT0000515 |
Mouse | mmu-miR-30d-3p | MIMAT0017011 |
Stem-loop Details
Mouse
