Assay Details
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | UGUAAACAUCCCCGACUGGAAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-30d-5p | MIMAT0000245 |
Mouse | mmu-miR-30d-5p | MIMAT0000515 |
Rat | rno-miR-30d-5p | MIMAT0000807 |
Anolis carolinensis | aca-miR-30d-5p | MIMAT0021926 |
Chicken | gga-miR-30d | MIMAT0001129 |
Fugu rubripes | fru-miR-30d | MIMAT0002949 |
View More Rows | ||
Gorilla gorilla | ggo-miR-30d | MIMAT0002618 |
Horse | eca-miR-30d | MIMAT0013006 |
Macaca nemestrina | mne-miR-30d | MIMAT0002619 |
Monodelphis domestica | mdo-miR-30e-5p | MIMAT0050218 |
Pan paniscus | ppa-miR-30d | MIMAT0002620 |
Pan troglodytes | ptr-miR-30d | MIMAT0002617 |
Pongo pygmaeus | ppy-miR-30d | MIMAT0015738 |
Rhesus monkey | mml-miR-30d-5p | MIMAT0006171 |
Tetraodon nigroviridis | tni-miR-30d | MIMAT0002950 |
Xenopus tropicalis | xtr-miR-30d | MIMAT0003657 |
Zebrafish | dre-miR-30d | MIMAT0001806 |
Stem-loop Details
Human
Mouse
Rat
Anolis carolinensis
Chicken
Fugu rubripes
Gorilla gorilla
Horse
Macaca nemestrina
Monodelphis domestica
Pan paniscus
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
Tetraodon nigroviridis
Xenopus tropicalis
Zebrafish
