Assay Details
Amplicon Length: | 87 |
Chromosome Location: | Chr. 8 - 134804570 - 134804570 [-] on 134804570 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-30d-3p | MIMAT0004551 |
Human | hsa-miR-30d-5p | MIMAT0000245 |
Stem-loop Details
Human
