Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSA™ ALTAIR™ 5 Multigas Detector Economy Kit
Featuring a 60 second span calibration and 10 second bump test, this detector is Galaxy™ System-compatible for easy calibration and record keeping
Specifications
Alarm Types | Audible, visual, vibrating |
---|---|
Battery Life | 15 hr. |
Dimensions (L x W x H) | 6.68 x 3.52 x 6.68 in. (16.96 x 8.94 x 16.96cm) |
Height (English) | 6.68 in. |
Height (Metric) | 16.96 cm |
Catalog Number | Mfr. No. | Resolution | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Resolution | Price | Quantity & Availability | |||||
18-039-318
|
MSA™
10094927 |
LEL: 1%, O2: 0.1%, CO: 1ppm, PH3: 0.1ppm | N/A | N/A | N/A | ||||
18-039-315
|
MSA™
10094924 |
LEL: 1%, O2: 0.1%, CO: 1ppm, H2S: 1ppm, NH3: 0.5ppm | N/A | N/A | N/A | ||||
18-039-317
|
MSA™
10094926 |
LEL: 1%, O2: 0.1%, CO: 1ppm, H2S: 1ppm, ClO2: 0.01ppm | N/A | N/A | N/A | ||||
18-039-314
|
MSA™
10094923 |
LEL: 1%, O2: 0.1%, CO: 1ppm, H2S: 1ppm, NO2: 0.1ppm | N/A | N/A | N/A | ||||
Description
- Thick rubber housing provides non-slip profile for secure grip, easily surviving 304.8cm (10 ft.) drop.
- MotionAlert™ feature activates when a unit does not detect movement for 30 seconds
- MSA exclusive instrument end of sensor life warning
- 40% less calibration gas used per minute than industry average
- Sensor response and clear times in under 15 seconds on LEL, O2, CO, H2S, and SO2 — 50% faster than industry average
- Greater signal stability and repeatability under changing or extreme
- High-resolution monochrome display screen
- Many e-chem sensor options
- Integral pump or diffusion option
- Rechargeable lithium-ion or alkaline battery options
- 17-language display capability
Economy Kits — Monochrome Display
- Chemical
- Construction
- Gas Detection
- General Industry
- Hazardous Materials
- Indoor Air Quality
- Manufacturing
- Nuclear
- Oil Gas
- Telecommunications
- Water/Wastewater
Certifications
- CE ATEX I I 2G
- Ex ia d IIC T4
- UL Class I, Division 1, ABCDFG, Class II, Class III
- CSA C22.2 No. 157 Class I, Div. 1, Groups ABCD
- ANZ Ex ia s I/IIC T4 (Zone 0)
- IEC Ex ia d e I/IIC T4
Warranty and Services
2 years under normal use conditions for LEL/O2/CO/H2S versions
Specifications
Audible, visual, vibrating | |
6.68 x 3.52 x 6.68 in. (16.96 x 8.94 x 16.96cm) | |
16.96 cm | |
6.68 in. | |
-4°F to +122°F | |
2 Years | |
520g | |
8.94 cm |
15 hr. | |
6.68 in. | |
65 | |
16.96 cm | |
-20°C to +50°C | |
18.4 oz. | |
3.52 in. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title