Learn More
BUCHI Receiving Flasks for Rotary Evaporators
SDP
Use these receiving flasks with any rotary evaporator. BUCHI Spherical Receiving Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for collecting condensed solvents.
$287.83 - $1462.00
Specifications
Type Receiving Flask
Shape Round
View More Specs
Products 13
Catalog Number Mfr. No. Volume (Metric) Material (Coating) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Material (Coating) Price Quantity & Availability  
05-405-005
SDP
View Documents
BUCHI
000421
50 mL Uncoated
Each for $364.42
Only null left
Add to Cart
05-405-006
SDP
View Documents
BUCHI
000422
100 mL Uncoated
Each for $287.83
Only null left
Add to Cart
05-405-007
SDP
View Documents
BUCHI
000423
250 mL Uncoated
Each for $289.16
Only null left
Add to Cart
05-405-008
SDP
View Documents
BUCHI
000424
500 mL Uncoated
Each for $293.00
Only null left
Add to Cart
05-405-058
SDP
View Documents
BUCHI
11060907
250 mL Plastic and Glass
Each for $578.01
Only null left
Add to Cart
05-402-655
SDP
View Documents
BUCHI
000425
1000 mL Uncoated
Each for $553.00
Only null left
Add to Cart
05-402-656
SDP
View Documents
BUCHI
000426
2000 mL Uncoated
Each for $1,327.00
Only null left
Add to Cart
05-402-657
SDP
View Documents
BUCHI
000427
3000 mL Uncoated
Each for $887.00
Only null left
Add to Cart
05-404-230
SDP
View Documents
BUCHI
025264
500 mL P+G
Each for $532.00
Only null left
Add to Cart
05-402-822
SDP
View Documents
BUCHI
020728
1000 mL P+G
Each for $792.00
Only null left
Add to Cart
05-402-533
SDP
View Documents
BUCHI
040776
2000 mL P+G LT
Each for $698.00
Only null left
Add to Cart
05-404-232
SDP
View Documents
BUCHI
025266
3000 mL P+G
Each for $1,144.00
Only null left
Add to Cart
05-404-231
SDP
View Documents
BUCHI
025265
2000 mL P+G
Each for $1,462.00
Only null left
Add to Cart
Description
Description
Use these receiving flasks with any rotary evaporator. Buchi™ Spherical Receiving Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for collecting condensed solvents.
  • Spherical Receiving Flasks for rotary evaporators with ball joint (35/20)
  • Exclusive use of DURAN borosilicate glass 3.3
  • High chemical resistance against acids, alkalis and organic substances
  • Resistant to thermal shocks and high temperatures combined with low thermal expansion
  • Available in volumes from 50 mL to 3000 mL
  • Available uncoated, with P+G coating or P+G LT for dry ice condensers
Specifications
Specifications
Receiving Flask
Round
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.