Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D1 Rabbit Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cyclin D1, Rabbit Monoclonal Antibody.

Specifications

Antigen Cyclin D1
Clone EPR2241IHC
Applications Immunohistochemistry
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Form Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Form Price Quantity & Availability  
RM2113R1
View Documents
Epredia
RM2113R1
1 mL Ready-to-Use N/A N/A N/A
RM2113R7
View Documents
Epredia
RM2113R7
7 mL Ready-to-Use N/A N/A N/A
RM2113RQ
View Documents
Epredia
RM2113RQ
12 mL Ready-to-Use N/A N/A N/A
Description

Description

Cyclin D1 or PRAD-1 or bcl-1 is a key cell cycle regulator. It functions in association with cdk4 and/or cdk6 by phosphorylating the retinoblastoma protein that releases transcription factors to initiate DNA replication. It is a putative proto-oncogene overexpressed in a wide variety of human neoplasms. In lymphoid lesions, with the exception of few rare diseases, it is specific for mantle cell lymphoma (MCL).

Host Species: Rabbit

Clone: EPR2241IHC

Isotype: IgG

Species Reactivity: Human. Others not tested.

Immunogen: A synthetic peptide corresponding to residues near the C-terminus of human Cyclin D1

Molecular Weight: 36 kDa

Positive Control: Mantle cell lymphoma

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Cyclin D1
Immunohistochemistry
Rabbit
Primary
EPR2241IHC
Unconjugated
A synthetic peptide corresponding to residues near the C-terminus of human Cyclin D1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.