Learn More
Epredia™ Lab Vision™ GRP94 Rabbit Polyclonal Antibody with BSA and azide
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ GRP94, Rabbit Polyclonal Antibody.
Specifications
Antigen | GRP94 |
---|---|
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
RB10642P0
|
Epredia™
RB10642P0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
RB10642P1
|
Epredia™
RB10642P1 |
Concentrated | 500 μL | N/A | N/A | N/A | |||
RB10642P
|
Epredia™
RB10642P |
Concentrated | 1 mL | N/A | N/A | N/A | |||
RB10642R1
|
Epredia™
RB10642R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
RB10642R7
|
Epredia™
RB10642R7 |
Prediluted | 7 mL | N/A | N/A | N/A | |||
Description
Heat shock protein (HSP) molecular chaperones are environmental stress inducible gene products. The human HSP 90 family consists of 17 genes classified into four classes: HSP90AA, HSP90AB, HSP90B and TRAP. HSP 90 family members guide the normal folding, intracellular disposition and proteolytic turnover of many key regulators of cell growth, differentiation and survival. GRP 94, glucose regulated protein 94 kDa, a HSP90 family member, (also known as tumor rejection antigen 1 (TRA1), ECGP and GP96), localizes to the ER, is highly expressed in BGC-823 human gastric carcinoma cells and is upregulated in human endothelial cells in response to hypoxia by HIF-1. TRAP1 (TNF receptor-associated protein 1), also designated HSP 75 is a mitochondrial matrix component that plays a role in the induction of apoptosis in response to reactive oxygen species.Host Species: Rabbit
Species Reactivity: Human. Others not known.
Epitope: C-terminal
Immunogen: Synthetic peptide corresponding to C-terminal of human GRP94
Molecular Weight: 94 kDa
Positive Control: Breast CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
GRP94 | |
Polyclonal | |
Rabbit | |
Cancer and Tumor Biology | |
Synthetic peptide corresponding to C-terminal of human GRP94 | |
Breast CA |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.