Learn More
Precipio™ HemeScreen MPN Panels
The HemeScreen MPN Panel-4 is an RUO pre-plated kit used to detect MPN-related gene mutations for JAK2, MPL, and CALR genes through HRM methods.
$1201.00 - $2410.00
Specifications
Content And Storage | 96-well Plate with Gene Targets, Positive, Negative, and Wild Type Controls. Store at -20°C |
---|---|
Detection Method | HRM |
For Use With (Equipment) | PCR Instruments |
Includes Label or Dye | Yes |
Sensitivity | 0.95 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
22-021-968
|
Precipio™
HS4PMPN |
4 Samples |
Each for $1,201.00
|
|
|||||
22-021-969
|
Precipio™
HS8PMPN |
8 Samples |
Each for $2,410.00
|
|
|||||
Description
HemeScreen is a proprietary set of RUO (Research Use Only) reagents used to screen the wild type (Negative) from Mutated (Positive) genes in a simplified workflow relative to alternative molecular testing technologies (RT-PCR or NGS).
- Pre-plated configuration with embedded mutant, wild type, and NTC controls to run 4 or 8 samples in duplicate
- Simplified workflow and reduced chance of error
- Data capture during thermocycling
- Fast turnaround time
- Broad mutation detection (SNPs, Modifications, Indels)
- Low limits of detection
- Results are qualitative and DNA sequencing is required to identify specific isoforms
- LOD: 5%
- Assay includes Primers-Mastermix Mix and controls
- Requires HRM-enabled RT-PCR (example Thermo Fisher™ Quantstudio 3 or higher)
Specifications
96-well Plate with Gene Targets, Positive, Negative, and Wild Type Controls. Store at -20°C | |
PCR Instruments | |
0.95 | |
JAK2 Ex. 14 (V617F), JAK2 Ex. 12, JAK2 Ex. 13, MPL, CALR |
HRM | |
Yes | |
Dry Ice | |
>99% |
Safety and Handling
ShelfLife : 1 Year
Your input is important to us. Please complete this form to provide feedback related to the content on this product.