Learn More
Biocartis Idylla™ Molecular Diagnostics Single-Use Cartridges
Idylla™ Single-Use Cartridges perform all processing steps in one cartridge.
Specifications
Quantity | 6 cartridges/box |
---|---|
For Use With (Application) | DNA Extraction, Real Time PCR (qPCR) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-555-100
|
Biocartis
A0011/6 |
6 cartridges/box | N/A | N/A | N/A | ||||
23-555-101
|
Biocartis
A0021/6 |
6 cartridges/box | N/A | N/A | N/A | ||||
23-555-102
|
Biocartis
A0031/6 |
6 cartridges/box | N/A | N/A | N/A | ||||
23-555-103
|
Biocartis
A0061/6 |
6 cartridges/box | N/A | N/A | N/A | ||||
23-555-104
|
Biocartis
A0071/6 |
6 cartridges/box | N/A | N/A | N/A | ||||
23-555-105
|
Biocartis
A0081/6 |
6 cartridges/box | N/A | N/A | N/A | ||||
23-555-106
|
Biocartis
A0091/6 |
6 cartridges/box | N/A | N/A | N/A | ||||
23-555-110
|
Biocartis
A0101/6 |
6 cartridges/box | N/A | N/A | N/A | ||||
Description
- Single-use disposable cartridges
- Polypropylene construction
- Contimaination-controlled design
- All reagents on-board
- Applicable for a wide variety of clinical samples
- All processing steps in one powerful cartridge
- Liquefaction
- Cell lysis
- DNA/RNA extraction
- Real-time PCR amplification and detection
- Data analysis and reporting
Specifications
6 cartridges/box | |
DNA Extraction, Real Time PCR (qPCR) |