Learn More
Epredia™ Lab Vision™ IgM (μ-Heavy Chain) Ab-2 Rabbit Polyclonal Antibody with BSA and azide
IgM (μ-Heavy Chain) Ab-2, Rabbit PAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Specifications
Antigen | IgM (µ-Heavy Chain) Ab-2 |
---|---|
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
RB1434A0
|
Epredia™
RB1434A0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
RB1434A1
|
Epredia™
RB1434A1 |
Concentrated | 500 μL | N/A | N/A | N/A | |||
RB1434A
|
Epredia™
RB1434A |
Concentrated | 1 mL | N/A | N/A | N/A | |||
RB1434R1
|
Epredia™
RB1434R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
RB1434R7
|
Epredia™
RB1434R7 |
Prediluted | 7 mL | N/A | N/A | N/A | |||
Description
The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not known.
Immunogen: IgM isolated from human serum of a patient with Waldenstrom's
Molecular Weight: 50kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic Ab-2 recognizes the μ heavy chain of human immunoglobulins. Ab-2 does not cross-react with γ (IgG), α (IgA), ε (IgE), or δ (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. Ab-2 is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
IgM (µ-Heavy Chain) Ab-2 | |
Polyclonal | |
Rabbit | |
Immunoglobulins | |
IgM isolated from human serum of a patient with Waldenstroms | |
Primary |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.