Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega BacTiter-Glo™ Microbial Cell Viability Assay Kit
Shop All Promega Corporation Products
Click to view available options
Quantity:
10 mL
100 mL
10 x 10mL
10 x 100mL
For Use With (Application):
Antimicrobial Compound Activity, Antimicrobial Compounds Screening, Viability Assay, Rapid Bacterial Quantitation, Rapid ATP Detection
Viability Assay
Description
Includes
BacTiter-Glo Buffer; BacTiter-Glo Substrate (lyophilized)
“Add-mix-measure” format utilizes a single reagent that causes cell lysis and generation of a luminescent signal after direct addition to bacterial cells cultured in medium. The luminescent signal is proportional to the amount of ATP present, which is directly proportional to the number of viable cells in culture.
- Generates a stable, “glow-type” luminescent signal with a half-life generally over 30 minutes
- Detects ATP from as few as 10 bacterial cells
- Produces results quickly—data can be recorded in five minutes or less after reagent addition
- Requires fewer handling steps than other ATP assays, making the kit ideal for automation
- Compatible for use with 96- or 384-well plates
- U.S. Patent 6,602,677, Australian Patent 754312 and other patents pending.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
Specifications
Specifications
Content And Storage | -20°C |
For Use With (Application) | Viability Assay |
For Use With (Equipment) | Microplate Reader |
Quantity | 10 x 100mL |
Sufficient For | 100,000 Assays at 100μL/assay in 96-well plates, 400,000 assays at 25μL/assay in 384-well plates |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction