Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Raptor™ HILIC-Si LC Columns, Particle Size: 2.7 μm
SDP

Catalog No. p-7199291 Shop All Restek Corporation Products
Click to view available options
Length (Metric):
30 mm
50 mm
100 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm

Designed for increased retention of polar compounds without ion-pairing reagents

  • 2.7 μm Raptor core-shell particles provide the speed of SPP, with greater efficiency and capacity than 5 μm particles
  • Fully compatible with both HPLC and UHPLC when using low back-pressure HILIC mobile phases
  • Switch to a Raptor HILIC-Si LC column when more retention and resolution of hydrophilic, polar analytes is needed
  • Ideal for increasing sensitivity and selectivity in LC-MS analyses

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 30 mm
Max. Pressure 600 bar (8,700 psi)
Particle Size 2.7 μm
pH 2 to 8
Pore Size 90 Å
Surface Area 130 m2/g
Temperature Max: 80°C
USP Type L13
Stationary Phase Bare Silica
Packing Material Superficially Porous Silica (SPP or Core-Shell)
Type Column
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.