Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Roc™ LC Guard Column Cartridges
Shop All Restek Corporation Products

Click to view available options
For Use With:
Roc Phenyl-Hexyl HPLC Columns
Roc Silica HPLC Columns
Roc™ C18 HPLC Columns
Roc™ C8 HPLC Columns
Roc™ Cyano HPLC Columns
Description
- Guard Column Cartridges require Roc™ Guard Column Holder (cat. #25812)
Specifications
Specifications
| Diameter (Metric) | 4 mm |
| For Use With | Roc™ C8 HPLC Columns |
| Length (Metric) | 10 mm |
| Quantity | 3/Pk. |
| Type | Guard Cartridge |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction