Learn More
3M™ SecureFit™ 400 Series Safety Glasses
3M™ SecureFit™ 400 Series Safety Glasses offer models with stylized colors, proprietary Pressure Diffusion Temple (PDT) technology for comfort, and a contoured brow guard to help provide additional coverage from flying particles.
$14.65 - $293.45
Specifications
Lens Material Polycarbonate
View More Specs
Products 15
Catalog Number Mfr. No. Frame Color Lens Tint LensType Price Quantity & Availability  
Catalog Number Mfr. No. Frame Color Lens Tint LensType Price Quantity & Availability  
19-070-944
View Documents
3M™
7100179972
Yellow, Black Amber
Case for $289.28
Only null left
Add to Cart
19-070-947
View Documents
3M™
7100180051
Blue and Gray Indoor/Outdoor
Case for $290.32
Only null left
Add to Cart
19-070-948
View Documents
3M™
7100180050
Blue and Gray Clear
Case for $290.32
Only null left
Add to Cart
19-070-949
View Documents
3M™
7100180049
Blue and Gray Clear
Case for $293.45
Only null left
Add to Cart
19-070-951
View Documents
3M™
7100180047
Black, Green Gray
Case for $205.83
Only null left
Add to Cart
19-070-952
View Documents
3M™
7100180045
Black, Green Clear
Case for $205.83
Only null left
Add to Cart
17-623-766
View Documents
3M™
7100112436
Black, Yellow Indoor/Outdoor Mirror
Each for $14.65
Case of 20 EA for $293.04
Only null left
Add to Cart
17-623-767
View Documents
3M™
7100112435
Black, Yellow Clear
Each for $14.65
Case of 20 EA for $293.04
Only null left
Add to Cart
17-623-768
View Documents
3M™
7100112433
Black, Yellow Gray
Each for $14.65
Case of 20 EA for $293.04
Only null left
Add to Cart
19-070-942
View Documents
3M™
7100180052
Black, Green Gray N/A N/A N/A
19-070-943
View Documents
3M™
7100180054
Pink, Black Gray N/A N/A N/A
19-070-945
View Documents
3M™
7100180036
Black, Gray Orange N/A N/A N/A
19-070-946
View Documents
3M™
7100179970
Light Blue, White Clear N/A N/A N/A
19-070-950
View Documents
3M™
7100180048
Hi-Vis Green and Clear Clear N/A N/A N/A
19-070-953
View Documents
3M™
7010528317
Black, Green Clear N/A N/A N/A
Description
Description
  • Contoured brow guard rests comfortably against forehead and helps provide additional coverage from flying particles
  • Features proprietary 3M™ Pressure Diffusion Temple Technology which helps diffuse pressure over the ear for a secure, comfortable fit
  • Enhanced temple comfort without compromising the security of fit across diverse workforces
  • Soft nosepiece to help reduce slippage
  • Applied on both sides of the lens, 3M™ Scotchgard™ Anti-fog is a long-lasting coating that provides better fog and scratch resistance when compared to traditional anti-fog coatings
  • 3M Catalog# SF401XAF-GRN, SF401XAS-GRN, SF401XSGAF-BLU, SF401XSGAF-GRN, SF401XSGAF-WHT, SF402XAF-GRN, SF402XAS-GRN, SF402XSGAF-BLU, SF403XSGAF-YEL, SF407XSGAF-BLU, SF409XAS-PNK, SF416XAS-BLK
Specifications
Specifications
Polycarbonate
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.