Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Squids™ 3790 Tool Shackle

Squids™ 3790 Tool Shackles connect to waisted/necked tools or tools with captive holes, creating clean and secure attachment points for tethering.

$13.00 - $29.50

Specifications

Certifications/Compliance Approved to the ANSI/ISEA 121-2018 standard
Includes 2/Pk.
Load Bearing Capacity (English) 15 lb.
Load Bearing Capacity (Metric) 6.8 kg
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-820-356
View Documents
Ergodyne™
19792
Small
Pack of 2 for $29.50
Only null left
Add to Cart
 
19-820-357
View Documents
Ergodyne™
19793
Medium
Pack of 2 for $13.00
Only null left
Add to Cart
 
19-820-358
View Documents
Ergodyne™
19794
Large
Pack of 2 for $13.50
Only null left
Add to Cart
 
19-820-359
View Documents
Ergodyne™
19795
X-Large
Pack of 2 for $21.50
Only null left
Add to Cart
 
Description

Description

  • Approved to the ANSI/ISEA 121-2018 standard
  • 304 stainless steel material
  • One step retrofit attachment for tethering point
  • Connects around midsection of a waisted or necked tool
  • Secure by adding a drop of adhesive and tightening
Specifications

Specifications

Approved to the ANSI/ISEA 121-2018 standard
2/Pk.
15 lb.
6.8 kg
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.