Learn More
Honeywell™ Uvex™ Seismic™ Sealed Eyewear
Eye protection against impact, sun, wind, dust and airborne debris
Catalog Number | Mfr. No. | Frame Color | Lens Coating | Lens Tint | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Frame Color | Lens Coating | Lens Tint | Price | Quantity & Availability | |||
19-351-460
|
Honeywell™
S0624X |
Metallic Blue | Anti-fog | SCT-Reflect 50 | N/A | N/A | N/A | ||
19-351-459
|
Honeywell™
S0622X |
Metallic Blue | Amber | N/A | N/A | N/A | |||
19-351-458
|
Honeywell™
S0620X |
Metallic Blue | Clear | N/A | N/A | N/A | |||
19-351-455
|
Honeywell™
S0602X |
Black | Anti-fog | Amber | N/A | N/A | N/A | ||
19-351-457
|
Honeywell™
S0620 |
Metallic Blue | Hardcoat | Clear | N/A | N/A | N/A | ||
19-351-456
|
Honeywell™
S0604X |
Black | Anti-fog | SCT-Reflect 50 | N/A | N/A | N/A | ||
19-351-454
|
Honeywell™
S0601X |
Black | Anti-fog | Espresso | N/A | N/A | N/A | ||
19-351-452
|
Honeywell™
S0600D |
Black | Anti-fog | Clear | N/A | N/A | N/A | ||
19-351-451
|
Honeywell™
S0600 |
Black | Clear | N/A | N/A | N/A | |||
19-185-650
|
Honeywell™
S0600HS |
Black | Anti-fog | Clear | N/A | N/A | N/A | ||
19-185-651
|
Honeywell™
S0601HS |
Black | Espresso | N/A | N/A | N/A | |||
Description
- For dirty, high-particulate work environments
- With Dura-streme dual coating technology which combines two types of lens coatings
- Uvextreme anti-fog (interior) and Ultra-dura anti-scratch (exterior), as well as Uvextra AF anti-fog/Hardcoat anti-scratch coating options
- Soft and flexible nasal pads
- Soft, high-quality face cushioning and padded temple tips
- Highly adjustable flame-resistant headband
- Replaceable flame-resistant, cushion-lined subframe
Applications: For applications like grinding, chipping, shaving and sanding.
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Removable |