The ValueWare Advantage
Manufactured from KIMAX™ borosilicate glass, ValueWare has a low coefficient of expansion and a very high resistance to chemical attack. This means optimal performance in the lab at an affordable price.
- Long life expectancy, resistant to most chemicals
- Ideal for general laboratory use
- Dependable, high quality
- Durable, sturdy wall thickness and excellent pouring characteristics
- Industry-standard packaging
- Economical