UserName
Viewing profile as user:
Solvents Resource Center
Solvents Resource Center
Common Uses of Solvents
Solvents are liquid, gas, or solid chemicals that dissolve, extract, or suspend other substances. The most common and universal solvent is water, which dissolves more substances than any other.
Solvents are often categorized as polar or nonpolar, a factor of the solvent’s structure and charge that determines the types of substances it can dissolve. Polar solvents have a positive and a negative charge at different places in their structures and will dissolve other polar substances. Water, acetone, acetonitrile, dimethylformamide (DMF), dimethyl sulfoxide (DMSO), ethanol, isopropanol, and methanol are examples of polar solvents.
Nonpolar solvents include alkanes (pentane, hexane, and heptane) and aromatics (benzene, toluene, and xylene). Other common nonpolar solvents include acetic acid, chloroform, diethyl ether, ethyl acetate, methylene chloride, and pyridine. Nonpolar organic solvents are used as spot removers, paint thinners, nail polish removers, glue solvents, and in perfumes and detergents.
Solvents are used extensively as a medium for chemical reactions and analytical separations and are available in various purity grades and packaging options. They are also essential for manufacturing cosmetics, pharmaceuticals, personal care items, textiles, paints, pesticides, and many other products.
Long-term exposure to some solvents can be hazardous to worker health, however. Chloroform and benzene, for example, are commonly found in gasoline and considered carcinogenic. Solvents can also damage the liver, kidneys, nervous system, or the brain. Chronic solvent-induced encephalopathy (CSE) is a known condition. Occupational exposure to organic solvents is associated with alcoholism, cataracts, and the loss of hearing and color vision.
Top Solvents
Learn about each solvent’s physical and chemical properties, uses, designations and grades, and more.
Top Solvents
Related Resources
Chemical and Chromatography Specialists
Our team is ready to answer your questions and help you select products.
Chemical and Chromatography Specialists
Get Solvents Shipped the Same Day
Find a range of solvents available for same-day shipping through the Fisher Scientific Edge program.
Fisher Scientific Edge program
Bulk and Custom Chemical Solutions
Our specialists can help you find what works best for your operation.
Bulk and Custom Chemical Solutions
Chemical Storage and Management Resource Center
Access safety data sheets, product certificates, and more.
Chemical Storage and Management Resource Center
Additional Solvents Resources
Download the following resources to learn more about solvent selection and browse our portfolio.
Returnable Solvent Containers Data Sheet
See how reusable solvent containers can help solve contamination problems and streamline your operations.
Returnable Solvent Containers Data Sheet
Solvents Application Guide
Learn more about Thermo Scientific and Fisher Chemical products, packaging, and delivery systems.
Solvents Application Guide
Fisher Chemical Solvents Data Sheet
Browse solvents suited for HPLC, gas chromatography, and more while reducing your chemical footprint.
Fisher Chemical Solvents Data Sheet
Honeywell Solvents Whitepaper
Review how Honeywell solvents perform compared to other leading brands.
Honeywell Solvents Whitepaper
More
Honeywell ACS Solvents Data Sheet
Discover reagent-grade solvents for general laboratory use, a value priced solution that meets your needs.
Chromatography Workflow Brochure
Find solvents for each step of your sample preparation and HPLC workflows.
Applications
Chromatography
Find solvents, reagents, and more for liquid and gas chromatography.
Chromatography
Cell Analysis Methods
Explore products for flow cytometry, cell imaging, and more.
Cell Analysis Methods
Protein Biology Methods
Browse isolation, purification, separation, and detection essentials.
Protein Biology Methods
Molecular Biology Methods
Explore products and resources for molecular biology protocols.
Molecular Biology Methods