Assay Details
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | CUUUCAGUCAGAUGUUUGCUGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-30d-3p | MIMAT0004551 | |
| Mouse | mmu-miR-30d-3p | MIMAT0017011 | |
| Rat | rno-miR-30d-3p | MIMAT0004722 | |
| Chrysemys picta | cpi-miR-30d-3p | MIMAT0037682 | |
| Columba livia | cli-miR-30d-3p | MIMAT0038457 | |
| Cricetulus griseus | cgr-miR-30d | MIMAT0023899 | |
| View More Rows | |||
| Dasypus novemcinctus | dno-miR-30d-3p | MIMAT0047564 | |
| Guinea pig | cpo-miR-30d-3p | MIMAT0046911 | |
| Monodelphis domestica | mdo-miR-30e-3p | MIMAT0050219 | |
| Pteropus alecto | pal-miR-30d-3p | MIMAT0039825 | |
| Rabbit | ocu-miR-30d-3p | MIMAT0048158 | |
Stem-loop Details
Human
Mouse
Rat
Chrysemys picta
Columba livia
Cricetulus griseus
Dasypus novemcinctus
Guinea pig
Monodelphis domestica
Pteropus alecto
Rabbit
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool B v3.0Megaplex™ RT Primers, Rodent Pool B v3.0
Associated TaqMan® Array MicroRNA Cards:
TaqMan® Human MicroRNA B Cards v3.0TaqMan® Rodent MicroRNA B Cards v3.0
TaqMan® OpenArray® MicroRNA Panels:
TaqMan® OpenArray® Human MicroRNA PanelTaqMan® OpenArray® Rodent MicroRNA Panel
Related Products
miRBase Alias:
Human: hsa-miR-30d* (v17)
Mouse: mmu-miR-30d* (v17)
Rat: rno-miR-30d* (v18)
Mouse: mmu-miR-30d* (v17)
Rat: rno-miR-30d* (v18)

Genomic Map