Assay Details
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | CUUUCAGUCAGAUGUUUGCUGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-30d-3p | MIMAT0004551 |
| Mouse | mmu-miR-30d-3p | MIMAT0017011 |
| Rat | rno-miR-30d-3p | MIMAT0004722 |
| Chrysemys picta | cpi-miR-30d-3p | MIMAT0037682 |
| Columba livia | cli-miR-30d-3p | MIMAT0038457 |
| Cricetulus griseus | cgr-miR-30d | MIMAT0023899 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-30d-3p | MIMAT0047564 |
| Guinea pig | cpo-miR-30d-3p | MIMAT0046911 |
| Monodelphis domestica | mdo-miR-30e-3p | MIMAT0050219 |
| Pteropus alecto | pal-miR-30d-3p | MIMAT0039825 |
| Rabbit | ocu-miR-30d-3p | MIMAT0048158 |
Stem-loop Details
Human
Mouse
Rat
Chrysemys picta
Columba livia
Cricetulus griseus
Dasypus novemcinctus
Guinea pig
Monodelphis domestica
Pteropus alecto
Rabbit

Genomic Map