Assay Details
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | CUUUCAGUCAGAUGUUUGCUGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-30d-3p | MIMAT0004551 |
Mouse | mmu-miR-30d-3p | MIMAT0017011 |
Rat | rno-miR-30d-3p | MIMAT0004722 |
Chrysemys picta | cpi-miR-30d-3p | MIMAT0037682 |
Columba livia | cli-miR-30d-3p | MIMAT0038457 |
Cricetulus griseus | cgr-miR-30d | MIMAT0023899 |
View More Rows | ||
Dasypus novemcinctus | dno-miR-30d-3p | MIMAT0047564 |
Guinea pig | cpo-miR-30d-3p | MIMAT0046911 |
Monodelphis domestica | mdo-miR-30e-3p | MIMAT0050219 |
Pteropus alecto | pal-miR-30d-3p | MIMAT0039825 |
Rabbit | ocu-miR-30d-3p | MIMAT0048158 |
Stem-loop Details
Human
Mouse
Rat
Chrysemys picta
Columba livia
Cricetulus griseus
Dasypus novemcinctus
Guinea pig
Monodelphis domestica
Pteropus alecto
Rabbit

More Information
Related Products
miRBase Alias:
Human: hsa-miR-30d* (v17)
Mouse: mmu-miR-30d* (v17)
Rat: rno-miR-30d* (v18)
Mouse: mmu-miR-30d* (v17)
Rat: rno-miR-30d* (v18)
