CAS RN 7689-03-4
CAS RN 7689-03-4
Thermo Scientific Chemicals (+)-Camptothecin, 98%
CAS: 7689-03-4 Molecular Formula: C20H16N2O4 Molecular Weight (g/mol): 348.35 MDL Number: MFCD00081076 InChI Key: VSJKWCGYPAHWDS-FQEVSTJZSA-N Synonym: camptothecin,camptothecine,s-+-camptothecin,campathecin,+-camptothecine,d-camptothecin,20 s-camptothecine,+-camptothecin,21,22-secocamptothecin-21-oic acid lactone,s-camptothecin PubChem CID: 24360 ChEBI: CHEBI:27656 SMILES: CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O
Thermo Scientific Chemicals Camptothecin
CAS: 7689-03-4 Synonym: (S)-(+)-Camptothecin
Tocris Bioscience™ Camptothecin
CAS: 7689-03-4 Molecular Formula: C20H16N2O4
MP Biomedicals, Inc Camptothecin 97% MP Biomedicals
CAS: 7689-03-4 Molecular Formula: C20H16N2O4 Molecular Weight (g/mol): 348.36 MDL Number: MFCD00081076 InChI Key: VSJKWCGYPAHWDS-FQEVSTJZSA-N Synonym: camptothecin,camptothecine,s-+-camptothecin,campathecin,+-camptothecine,d-camptothecin,20 s-camptothecine,+-camptothecin,21,22-secocamptothecin-21-oic acid lactone,s-camptothecin PubChem CID: 24360 ChEBI: CHEBI:27656 SMILES: CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O
(S)-(+)-Camptothecin 97.0+%, TCI America™
CAS: 7689-03-4 Molecular Formula: C20H16N2O4 Molecular Weight (g/mol): 348.36 MDL Number: MFCD00081076 InChI Key: VSJKWCGYPAHWDS-FQEVSTJZSA-N Synonym: camptothecin,camptothecine,s-+-camptothecin,campathecin,+-camptothecine,d-camptothecin,20 s-camptothecine,+-camptothecin,21,22-secocamptothecin-21-oic acid lactone,s-camptothecin PubChem CID: 24360 ChEBI: CHEBI:27656 SMILES: CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O