Diagnostic Equipment
ClinicDx   
   Bringing Hospital Lab Expertise to Your Clinic
ClinicDx   
Bringing Hospital Lab Expertise to Your Clinic
Explore centrifuges, refrigerators, freezers, incubators, laboratory seating, and other products to support your work, all selected for ClinicDx and supported by the expertise of the Fisher Healthcare team.
Refrigerators and freezers are available in many configurations, including undercounter, countertop, and full-sized units. Refrigerators may also be designed specifically for storing vaccines or other materials with special requirements. Choose countertop or full-sized -20°C freezers for sample and reagent storage.
Centrifuges are available with multiple options for speed and size and may be designed for specific rotors or sample types. Some centrifuges have single or multiple preset speeds for easier repeat use. Very small “mini,” compact, and higher-capacity units are available with and without refrigeration. Rotors may be fixed angle, horizontal, or both. Some centrifuges are specifically designed for processing microhematocrits, performing blood sample preparations, or spinning urine.
Incubator options include basic or programmable microbiological or CO2 incubators. Other benchtop wet and dry baths in a range of sizes are also available.
Microscopes may be analog or digital, and specifically configured for routine microscopy, hematology, cytopathology, microbiology, histology, or other clinical applications. Digital capabilities make it easier to capture, catalog, and share images. Choose brightfield, darkfield (fluorescence), polarization, phase contrast, or other illumination methods.
Laboratory seating options include stools and chairs in a range of adjustable or fixed heights, and vinyl seat covers in multiple color options.
Count on ClinicDx, supported by experts from the Fisher Healthcare team, as your trusted resource to help you quickly find the high-quality diagnostic equipment you need.
  More
Filtered Search Results
Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.
Narrow Results
Narrow Results
  • (6)
  • (6)
  • (2)
  • (4)
  • (15)