Firefighting and Rescue Gloves
- (18)
- (1)
- (15)
- (40)
- (10)
- (3)
- (5)
- (1)
- (1)
- (15)
- (11)
- (14)
- (10)
- (3)
- (8)
- (15)
- (14)
- (10)
- (6)
- (1)
- (5)
- (12)
- (8)
- (9)
- (40)
- (6)
- (1)
- (1)
- (6)
- (4)
- (5)
- (10)
- (1)
- (3)
- (7)
- (10)
Filtered Search Results
Honeywell™ Kangaroo Leather Super Glove™ - Gauntlet Cuff
3D hand-shaped design with staggered layer seaming is ergonomically correct, helping to fight hand fatigue and reduce injuries. Digiroo™ High Grip technology on the palm and fingers ensures dexterity, grip, and tactility.
| Includes | Crosstech™ Moisture Barrier |
|---|---|
| Series | GL-SGKCG |
| Product Type | Structural Firefighting Gloves |
| Back Material | Top Grain Kangaroo Leather |
| Color | Blue |
| Thumb Style | Wing |
| Coated Area | Palm, Fingers |
| Certifications/Compliance | NFPA 1971-2007 |
| Liner Material | Nomex™/Aramid Fiber |
| Cuff Material | Cowhide Leather |
| Glove Cuff Style | Gauntlet |
| Hand | Specific |
| Glove Cuff Color | Black |
Honeywell™ Eclipse™ 8700 Structural Firefighting Gloves
Innovative cuff design is convertible and can be worn as a knit wrist, or in a gauntlet style. DuPont™ Nomex™ fibers provide strength and durability, in addition to a high level of thermal protection.
| Includes | Pyrotect™ Moisture Barrier |
|---|---|
| Series | GL-8700 Eclipse |
| Product Type | Structural Firefighting Gloves |
| Coating Material | Uncoated |
| Back Material | Elkhide Leather |
| Color | Dark Blue,Yellow |
| Palm Type | Kangaroo Leather |
| Certifications/Compliance | NFPA 1971-2007 |
| Liner Material | Nomex™/Aramid Fiber |
| Cuff Material | Nomex™ Spandex |
| Glove Cuff Style | Gauntlet |
| Hand | Specific |
| Glove Cuff Color | White |
Ringers Rope Rescue Gloves
Suited for all rope handling applications
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
| Product Type | Extrication Gloves |
|---|---|
| Material | Neoprene,Synthetic Leather |
| For Use With (Application) | Rescue, Rappelling, and SWAT Situations |
| Glove Cuff Style | Hook and Loop |
Ringers Extrication Hybrid Gloves
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
| Product Type | Extrication Gloves |
|---|---|
| Color | Green |
| Cut Resistance Rating | ANSI A3 |
| For Use With (Application) | Firemen, Rescue Workers, Military, and Law Enforcement |
Honeywell™ Eclipse™ 6400 Structural Firefighting Gloves
Innovative cuff design is convertible and can be worn as a knit wrist, or in a gauntlet style. DuPont™ Nomex™ fibers provide strength and durability, in addition to a high level of thermal protection.
| Includes | Crosstech™ Moisture Barrier |
|---|---|
| Series | GL-6400 Eclipse |
| Product Type | Structural Firefighting Gloves |
| Coating Material | Uncoated |
| Back Material | Elkhide Leather |
| Color | Brown,Dark Blue |
| Palm Type | Cowhide Leather |
| Certifications/Compliance | NFPA 1971 |
| Liner Material | Self Extinguishing Fleece (SEF) |
| Cuff Material | Nomex™ Spandex |
| Glove Cuff Style | Gauntlet |
| Hand | Specific |
| Glove Cuff Color | White |
Honeywell™ 9550 Fire Mate™ Kangaroo Leather Firefighter Gloves
Kangaroo leather on the palm absorbs significantly less water than traditional leather, while also providing a high level of abrasion resistance.
| Includes | Crosstech™ Moisture Barrier |
|---|---|
| Series | Fire Mate™ 9550 |
| Product Type | Structural Firefighting Gloves |
| Coating Material | Uncoated |
| Back Material | Elkhide Leather |
| Palm Type | Kangaroo Leather |
| Thumb Style | Wing |
| Certifications/Compliance | NFPA 1971-2007,NFPA 1951-2007 |
| Liner Material | Nomex™/Aramid Fiber |
| Cuff Material | Leather |
| Glove Cuff Style | Gauntlet |
| Hand | Specific |
Honeywell™ Proximity Firefighting Gloves - Knit Wrist
Knit PBI and Aramid Fiber based aluminized material is the preferred shell for proximity turnout gear due to its flexibility, durability, and radiant heat reflectivity.
| Finger Style | Full |
|---|---|
| Coating Material | Uncoated |
| Color | Brown,Silver |
| Palm Type | Elkhide Leather |
| Thumb Style | Wing |
| Cuff Material | Leather Pull Tab |
| Hand | Specific |
| Glove Cuff Color | Gray |
| Series | GL-BPR-RWA |
| Product Type | Proximity Firefighting Gloves |
| Back Material | Aluminized PBI/Aramid Fiber |
| Certifications/Compliance | NFPA 1971-2007 |
| Liner Material | Nomex™/Aramid Fiber |
| Glove Cuff Style | Knit |
Honeywell™ Kangaroo Leather Super Glove™ - Knit Wrist
3D hand-shaped design with staggered layer seaming is ergonomically correct, helping to fight hand fatigue and reduce injuries. Digiroo™ High Grip technology on the palm and fingers ensures dexterity, grip, and tactility.
| Includes | Crosstech™ Moisture Barrier |
|---|---|
| Series | GL-SGKCW |
| Product Type | Structural Firefighting Gloves |
| Back Material | Top Grain Kangaroo Leather |
| Color | Blue |
| Thumb Style | Wing |
| Coated Area | Palm, Fingers |
| Certifications/Compliance | NFPA 1971-2007,NFPA 1951,NFPA 1999,NFPA 1992 |
| Liner Material | Nomex™/Aramid Fiber |
| Cuff Material | Nomex™ |
| Glove Cuff Style | Knit |
| Hand | Specific |
| Glove Cuff Color | Black |
Wells Lamont Industrial™ Y7112 High Visibility Jersey Glove
Hi-vis orange jersey glove with microdots on palm for grip. Knit wrist to keep debris out.
Honeywell™ Proximity Firefighting Gloves - Gauntlet Cuff
Knit PBI and Aramid Fiber based aluminized material is the preferred shell for proximity turnout gear due to its flexibility, durability, and radiant heat reflectivity.
| Series | GL-BPR-RGA |
|---|---|
| Product Type | Proximity Firefighting Gloves |
| Coating Material | Uncoated |
| Back Material | Aluminized PBI/Aramid Fiber |
| Color | Brown,Silver |
| Palm Type | Elkhide Leather |
| Thumb Style | Wing |
| Certifications/Compliance | NFPA 1971-2007 |
| Liner Material | Nomex™/Aramid Fiber |
| Glove Cuff Style | Gauntlet |
| Hand | Specific |
| Glove Cuff Color | Silver |
Shelby™ Xtrication™ Gloves
Anatomically designed for improved hand and finger dexterity
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
| Product Type | Extrication Gloves |
|---|---|
| Color | Black |
| Material | Cordura Nylon |
| Certifications/Compliance | CE PPE, EN 659:1996, EN 388:1994 |
| Liner Material | Fluffed Terry Cotton |
Shelby™ FDP™ Leather Gloves
Fire-retardant and heat-resistant heavyweight gloves meet the demands of firefighting
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
| Product Type | Structural Firefighting Gloves |
|---|---|
| Color | Tan |
| Material | Cowhide Leather,Pigskin Leather |
| Certifications/Compliance | NFPA 1971, CE PPE, EN 659:1996, EN 388:1994 |
| Liner Material | Thermal |
| Glove Cuff Style | Gauntlet |
Shelby™ Firewall Steamblock™ Gloves
Shipboard firefighting gloves protect the back of the hand from steam
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
| Product Type | Structural Firefighting Gloves |
|---|---|
| Material | Cowhide Leather |
| Certifications/Compliance | NFPA 1971, CE PPE, EN 659:1996, EN 388:1994 |
| Liner Material | Thermal |
| Glove Cuff Style | Knit |
Shelby™ Structural Direct Grip Gloves
Innovative wash-and-wear palm stays soft and supple even after repeated washing
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Shelby™ 5284 Flex Structural Fire Gloves
Durable koala tanned leather on the palm ensures ultra-soft, abrasion resistant protection. Insular textile construction on the back of the hand provides thermal and flame resistance without sacrificing flexibility.
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More