Assay Details
Gene Symbol: FNR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084097.1 | NP_001077566.1 | 4 - 5 | 702 | 77 | |
NM_001332422.1 | NP_001319047.1 | 3 - 4 | 1037 | 77 | ||
NM_101857.6 | NP_173431.1 | 4 - 5 | 794 | 77 | ||
GenBank mRNA | AK318722.1 | - | 3 - 4 | 924 | 77 | |
AY062739.1 | - | 4 - 5 | 586 | 77 | ||
AY114663.1 | - | 4 - 5 | 568 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
