Assay Details
| Gene Symbol: AT2G18465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179646.4 | NP_849977.1 | 4 - 5 | 511 | 78 | |
| GenBank mRNA | AY136462.1 | - | 4 - 5 | 412 | 78 | |
| BT001155.1 | - | 4 - 5 | 387 | 78 | ||
More Information
Set Membership: |
|

AT2G18465
Genomic Map