Assay Details
Gene Symbol: HB25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125939.3 | NP_201344.1 | 1 | 931 | 88 | |
GenBank mRNA | BT002385.1 | - | 1 | 839 | 88 | |
BT006304.1 | - | 1 | 754 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
