Assay Details
Gene Symbol: ARF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102198.5 | NP_564195.1 | 6 - 7 | 770 | 66 | |
GenBank mRNA | AF324684.2 | - | 6 - 7 | 659 | 66 | |
AF325025.2 | - | 6 - 7 | 659 | 66 | ||
AF329504.1 | - | 6 - 7 | 659 | 66 | ||
AF372875.1 | - | 6 - 7 | 659 | 66 | ||
AF372901.1 | - | 6 - 7 | 659 | 66 | ||
AY113940.1 | - | 5 - 6 | 543 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
