Assay Details
Gene Symbol: ras-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064222.7 | NP_496623.1 | 1 - 2 | 206 | 66 | |
NR_051787.1 | 1 - 2 | 113 | 66 | |||
GenBank mRNA | U56930.1 | - | 1 - 2 | 181 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
