Assay Details
Gene Symbol: pan-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065523.8 | NP_497924.2 | 3 - 4 | 437 | 93 | |
NM_065524.5 | NP_497925.1 | 8 - 9 | 1379 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
