Assay Details
| Gene Symbol: ddo-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066052.5 | NP_498453.1 | 2 - 3 | 178 | 108 | |
| GenBank mRNA | AB275893.1 | - | 2 - 3 | 145 | 108 | |
More Information
Set Membership: |
|

ddo-3
Genomic Map