Assay Details
Gene Symbol: ehbp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073067.4 | NP_505468.1 | 2 - 3 | 143 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
