Assay Details
| Gene Symbol: fib-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074290.6 | NP_506691.1 | 5 - 6 | 907 | 82 | |
| Gene Symbol: rps-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380824.2 | NP_001366849.1 | 4 - 5 | 951 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380824.2 transcripts.
Set Membership: |
|

fib-1
rps-16
Genomic Map