Assay Details
| Gene Symbol: ZNF410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623643.3 | XP_005623700.1 | 6 - 7 | 909 | 69 | |
| XM_005623644.3 | XP_005623701.1 | 6 - 7 | 812 | 69 | ||
| XM_022422206.1 | XP_022277914.1 | 6 - 7 | 815 | 69 | ||
| XM_862861.5 | XP_867954.2 | 6 - 7 | 909 | 69 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ZNF410
Genomic Map