Assay Details
Gene Symbol: TPD52L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003433182.4 | XP_003433230.1 | 2 - 3 | 292 | 65 | |
XM_003433183.4 | XP_003433231.1 | 2 - 3 | 292 | 65 | ||
XM_005635309.3 | XP_005635366.1 | 2 - 3 | 292 | 65 | ||
XM_022408988.1 | XP_022264696.1 | 2 - 3 | 1009 | 65 | ||
XM_022408989.1 | XP_022264697.1 | 2 - 3 | 292 | 65 | ||
XM_022408990.1 | XP_022264698.1 | 2 - 3 | 1020 | 65 | ||
XM_022408991.1 | XP_022264699.1 | 2 - 3 | 1568 | 65 | ||
XM_022408992.1 | XP_022264700.1 | 2 - 3 | 1016 | 65 | ||
XM_022408993.1 | XP_022264701.1 | 2 - 3 | 999 | 65 | ||
XM_022408994.1 | XP_022264702.1 | 2 - 3 | 999 | 65 | ||
XM_022408995.1 | XP_022264703.1 | 2 - 3 | 1012 | 65 | ||
XM_022408996.1 | XP_022264704.1 | 2 - 3 | 292 | 65 | ||
XM_022408997.1 | XP_022264705.1 | 2 - 3 | 1554 | 65 | ||
XM_022408998.1 | XP_022264706.1 | 2 - 3 | 981 | 65 | ||
XM_022408999.1 | XP_022264707.1 | 2 - 3 | 1004 | 65 | ||
XM_022409000.1 | XP_022264708.1 | 2 - 3 | 1023 | 65 | ||
XM_022409001.1 | XP_022264709.1 | 2 - 3 | 292 | 65 | ||
XM_022409002.1 | XP_022264710.1 | 2 - 3 | 1003 | 65 | ||
XM_022409003.1 | XP_022264711.1 | 2 - 3 | 1001 | 65 | ||
XM_848503.5 | XP_853596.1 | 2 - 3 | 291 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
